4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Diaminostilbene-2,2'-disulfonic acid, 95%, Thermo Scientific Chemicals
CAS: 81-11-8 | C14H14N2O6S2 | 370.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA406411000
Price (MYR)
359.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
359.00
Add to cart
Chemical Identifiers
CAS132-98-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white