Dextran, High Fraction, Thermo Scientific Chemicals
Dextran, High Fraction, Thermo Scientific Chemicals
Dextran, High Fraction, Thermo Scientific Chemicals
Dextran, High Fraction, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dextran, High Fraction, Thermo Scientific Chemicals
CAS: 9004-54-0 | (C6H9O5)n(C6H10O5)m | 504.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA406261000
Price (MYR)
858.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
858.00
Add to cart
Chemical Identifiers
CAS3970-35-2
IUPAC Name2-chloro-3-nitrobenzoic acid
Molecular FormulaC7H4ClNO4
InChI KeyJRQDVRIQJJPHEQ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(=C1Cl)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (Silylated GC)≥97.5%
FormPowder
Melting Point (clear melt)181-187?C
Appearance (Color)White to yellow