Cyclohexyldiphenylphosphine, 98%, Thermo Scientific Chemicals
Cyclohexyldiphenylphosphine, 98%, Thermo Scientific Chemicals
Cyclohexyldiphenylphosphine, 98%, Thermo Scientific Chemicals
Cyclohexyldiphenylphosphine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexyldiphenylphosphine, 98%, Thermo Scientific Chemicals
CAS: 6372-42-5 | C18H21P | 268.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA406100050
Price (MYR)
1,158.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,158.00
Add to cart
Chemical Identifiers
CAS108-12-3
IUPAC Name3-methylbutanoyl chloride
Molecular FormulaC5H9ClO
InChI KeyISULZYQDGYXDFW-UHFFFAOYSA-N
SMILESCC(C)CC(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.4135-1.4175 @ 20?C
Appearance (Color)Clear colorless