Cyclohexanone, 99+%, Thermo Scientific Chemicals
Cyclohexanone, 99+%, Thermo Scientific Chemicals
Cyclohexanone, 99+%, Thermo Scientific Chemicals
Cyclohexanone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexanone, 99+%, Thermo Scientific Chemicals
CAS: 108-94-1 | C6H10O | 98.145 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Catalog number FSA406092500
Price (MYR)
289.00
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (MYR)
289.00
Add to cart
Chemical Identifiers
CAS446-52-6
IUPAC Name2-fluorobenzaldehyde
Molecular FormulaC7H5FO
InChI KeyZWDVQMVZZYIAHO-UHFFFAOYSA-N
SMILESFC1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤2.25%
Refractive Index1.5195-1.5245 @ 20?C
Identification (FTIR)Conforms
FormLiquid
Appearance (Color)Clear colorless to yellow