β-Cyclodextrin, 98%, Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
CAS: 7585-39-9 | C42H70O35 | 1134.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number FSA406001000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Materialβ-Cyclodextrin
CAS7585-39-9
Melting Point310.0°C
EINECS Number231-493-2
Loss on Drying14% max. (105°C, 8 hrs)