β-Cyclodextrin, 98%, Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
β-Cyclodextrin, 98%, Thermo Scientific Chemicals
CAS: 7585-39-9 | C42H70O35 | 1134.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number FSA406000250
Price (MYR)
340.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
340.00
EA
Add to cart
Chemical Identifiers
CAS1205-91-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (GC)≥97.5%
Melting Point (clear melt)117.0-124.0?C
FormCrystals or powder or crystalline powder