Cyanocobalamin, 96%
Cyanocobalamin, 96%
Cyanocobalamin, 96%
Thermo Scientific Chemicals
Cyanocobalamin, 96%
CAS: 68-19-9 | C63H88CoN14O14P | 1355.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA405925000
Price (MYR)
550.00
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
550.00
Add to cart
Chemical Identifiers
CAS112-72-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormGranules, prills or crystalline powder
Assay (GC)>97.0%