Cyanocobalamin, 96%
Cyanocobalamin, 96%
Cyanocobalamin, 96%
Thermo Scientific Chemicals
Cyanocobalamin, 96%
CAS: 68-19-9 | C63H88CoN14O14P | 1355.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 mg
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA405920250
Price (MYR)
7,938.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
7,938.00
Add to cart
Specifications
Chemical Name or MaterialCyanocobalamin
Name Note96%
CAS68-19-9
Absorbance3.15 to 3.45 (A361/A(547 to 559)), 1.7 to 1.9 (A361/A278)
ColorRed