Cyanocobalamin, 96%
Cyanocobalamin, 96%
Cyanocobalamin, 96%
Thermo Scientific Chemicals
Cyanocobalamin, 96%
CAS: 68-19-9 | C63H88CoN14O14P | 1355.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
500 mg
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA405920010
Price (MYR)
756.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
756.00
Add to cart
Chemical Identifiers
CAS33286-22-5
IUPAC Namehydrogen (2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl acetate chloride
Molecular FormulaC22H27ClN2O4S
InChI KeyHDRXZJPWHTXQRI-BHDTVMLSSA-N
SMILES[H+].[Cl-].COC1=CC=C(C=C1)[C@@H]1SC2=CC=CC=C2N(CCN(C)C)C(=O)[C@@H]1OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Specific optical rotation+110° to +120° (20°C, 589 nm) (c=1, H2O)
Appearance (Form)Powder
HPLC>=97.5 %
Appearance (Color)White to off-white