Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper(II) nitrate, hemipentahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
CAS: 19004-19-4 | 2·5 H2O | 232.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA405855000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCopper(II) nitrate, hemipentahydrate
Melting Point114.0°C
CAS19004-19-4
ColorBlue
Health Hazard 1GHS Signal Word: Danger