Crystal Violet, certified
Crystal Violet, certified
Crystal Violet, certified
Crystal Violet, certified
Thermo Scientific Chemicals
Crystal Violet, certified
CAS: 548-62-9 | C25H30ClN3 | 407.986 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number FSA405835000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS15244-10-7
IUPAC Namediiron(3+) hydrate trisulfate
Molecular FormulaFe2H2O13S3
InChI KeyVXWSFRMTBJZULV-UHFFFAOYSA-H
SMILESO.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)73% Fe₂(SO₄)₃ min
Indicator
RUO – Research Use Only