p-Cresol, 99+%, pure, Thermo Scientific Chemicals
p-Cresol, 99+%, pure, Thermo Scientific Chemicals
p-Cresol, 99+%, pure, Thermo Scientific Chemicals
p-Cresol, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Cresol, 99+%, pure, Thermo Scientific Chemicals
CAS: 106-44-5 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number FSA405741000
Price (MYR)
254.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
254.00
Add to cart
Chemical Identifiers
CAS631-61-8
IUPAC Nameammonium acetate
Molecular FormulaC2H7NO2
InChI KeyUSFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES[NH4+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
pH6.5 to 7.5 (5% soln.in water)
UVat 250 nm A: =<0.05
UVat 270 nm A: =<0.03
UV(10 % in water)