Coumarin 2, 99%, laser grade
Coumarin 2, 99%, laser grade
Coumarin 2, 99%, laser grade
Coumarin 2, 99%, laser grade
Thermo Scientific Chemicals
Coumarin 2, 99%, laser grade
CAS: 26078-25-1 | C13H15NO2 | 217.268 g/mol
Have Questions?
Catalog NumberQuantity
FSA405485000500 mg
Catalog number FSA405485000
Price (MYR)
551.00
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (MYR)
551.00
Add to cart
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentGold, Au: ≤20ppm
Impurity contentPalladium, Pd: ≤200ppm
Impurity contentRuthenium, Ru: ≤20ppm
Impurity contentSodium, Na: ≤100ppm
Assay (unspecified)Loss On Reduction 1.9-3.5 wt%