2-Chloronicotinic acid, 99%
2-Chloronicotinic acid, 99%
2-Chloronicotinic acid, 99%
2-Chloronicotinic acid, 99%
Thermo Scientific Chemicals
2-Chloronicotinic acid, 99%
CAS: 2942-59-8 | C6H4ClNO2 | 157.56 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA40486050050 gGlass bottle
Catalog number FSA404860500
Price (MYR)
676.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
676.00
Add to cart
Specifications
Chemical Name or Material2-Chloronicotinic acid
Name Note98%
CAS2942-59-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning