2-Chloronaphthalene, 99%, Thermo Scientific Chemicals
2-Chloronaphthalene, 99%, Thermo Scientific Chemicals
2-Chloronaphthalene, 99%, Thermo Scientific Chemicals
2-Chloronaphthalene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Chloronaphthalene, 99%, Thermo Scientific Chemicals
CAS: 91-58-7 | C10H7Cl | 162.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA404840050
Price (MYR)
1,672.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,672.00
Add to cart
Specifications
Chemical Name or Material2-Chloronaphthalene
Name Note99%
CAS91-58-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning