Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA404415000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS50926-11-9
IUPAC Namestannanone indium
Molecular FormulaInOSn
InChI KeyAMGQUBHHOARCQH-UHFFFAOYSA-N
SMILES[In].O=[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Surface Area10-35m²/g
Assay (unspecified)99.5% % In₂O₃/SnO₂ min