Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroacetic acid sodium salt, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
500 g
Packaging:
View all
Glass Bottle
Catalog number FSA404410025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS163105-89-3
IUPAC Name(6-methoxypyridin-3-yl)boronic acid
Molecular FormulaC6H8BNO3
InChI KeyDHADXDMPEUWEAS-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=N1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥94.0%
Proton NMRConforms to structure