Cellulose Acetate Butyrate, Butyryl Content 35-39%
Cellulose Acetate Butyrate, Butyryl Content 35-39%
Cellulose Acetate Butyrate, Butyryl Content 35-39%
Cellulose Acetate Butyrate, Butyryl Content 35-39%
Thermo Scientific Chemicals
Cellulose Acetate Butyrate, Butyryl Content 35-39%
CAS: 9004-36-8 | (C12H14O10)n
Have Questions?
Catalog NumberQuantityPackaging
FSA404255000500 gPlastic bottle
Catalog number FSA404255000
Price (MYR)
739.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
739.00
Add to cart
Chemical Identifiers
CAS644-13-3
IUPAC Name(naphthalen-2-yl)(phenyl)methanone
Molecular FormulaC17H12O
InChI KeySJNXJRVDSTZUFB-UHFFFAOYSA-N
SMILESO=C(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)77-85?C
Appearance (Color)White to pale yellow or pale cream
Assay (GC)≥97.5%