Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
500 g
1 kg
Catalog number FSA404160025
Price (MYR)
828.00
Add to cart
Quantity:
2.5 kg
Request bulk or custom format
Price (MYR)
828.00
Add to cart
Chemical Identifiers
CAS112883-29-1
IUPAC Name(2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-hydroxyphenyl)propanoic acid
Molecular FormulaC24H21NO5
InChI KeySWZCTMTWRHEBIN-ANBDAQEENA-N
SMILESOC(=O)[C@@H](CC1=CC=C(O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%