Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Activated charcoal, NORIT(R) CN 1, decolorizing, Thermo Scientific Chemicals
CAS: 7440-44-0 | C | 12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
Packaging:
Glass bottle
Catalog number FSA404042500
Price (MYR)
293.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
293.00
Add to cart
Chemical Identifiers
CAS515-69-5
IUPAC Name(2S)-6-methyl-2-[(1R)-4-methylcyclohex-3-en-1-yl]hept-5-en-2-ol
Molecular FormulaC15H26O
InChI KeyRGZSQWQPBWRIAQ-GJZGRUSLSA-N
SMILESCC(C)=CCC[C@](C)(O)[C@@H]1CCC(C)=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4925-1.4985 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid or viscous liquid
Assay (GC)≥95.0%
Filter aid
RUO – Research Use Only