Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
CAS: 5794-28-5 | C2H2CaO5 | 146.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
Packaging:
Glass bottle
Catalog number FSA403885000
Price (MYR)
572.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
572.00
Add to cart
Specifications
Chemical Name or MaterialCalcium oxalate monohydrate
Melting Point200.0°C
Name Note98%
CAS5794-28-5