Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
CAS: 5794-28-5 | C2H2CaO5 | 146.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
Packaging:
Glass bottle
Catalog number FSA403880050
Price (MYR)
250.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
250.00
Add to cart
Chemical Identifiers
CAS81-84-5
SpecificationsSpecification SheetSpecification Sheet
Melting point>=269°C
Loss on drying=<0.5 %
Appearance (Color)Beige to light brown
Infrared spectrumConforms
HPLC>=96.0 %