Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA403575000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1309-64-4
IUPAC Namediantimony(3+) trioxidandiide
Molecular FormulaO3Sb2
InChI KeyGHPGOEFPKIHBNM-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Sb+3].[Sb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Iron (Fe)=<30 ppm
Appearance (Form)Powder
Arsenic (As)=<500 ppm
Lead (Pb)=<400 ppm