Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA403571000
Price (MYR)
553.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
553.00
Add to cart
Chemical Identifiers
CAS109511-58-2
IUPAC Name(2Z,3Z)-bis({amino[(2-aminophenyl)sulfanyl]methylidene})butanedinitrile
Molecular FormulaC18H16N6S2
InChI KeyDVEXZJFMOKTQEZ-JYFOCSDGSA-N
SMILESN\C(SC1=CC=CC=C1N)=C(\C#N)/C(/C#N)=C(\N)SC1=CC=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.0% (TLC)
Appearance (Color)White to off-white
Assay from Supplier's CofA≥99.0% (HPLC)
FormPowder or crystals or crystalline powder or granules
Melting Point130.0-133.0°C