Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Alfa Aesar
Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number FSA403570025
Price (MYR)
1,427.76
Add to cart
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,427.76
Add to cart
Specifications
Chemical Name or MaterialButyl 4-hydroxybenzoate
Name Note99+%
CAS94-26-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement