Bromothymol Blue sodium salt, ACS reagent
Bromothymol Blue sodium salt, ACS reagent
Bromothymol Blue sodium salt, ACS reagent
Bromothymol Blue sodium salt, ACS reagent
Thermo Scientific Chemicals
Bromothymol Blue sodium salt, ACS reagent
CAS: 34722-90-2 | C27H27Br2NaO5S | 646.37 g/mol
Have Questions?
Catalog NumberQuantity
FSA40326010010 g
Catalog number FSA403260100
Price (MYR)
387.00
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
387.00
Add to cart
Chemical Identifiers
CAS7440-48-4
IUPAC Namecobalt
Molecular FormulaCo
InChI KeyGUTLYIVDDKVIGB-UHFFFAOYSA-N
SMILES[Co]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
FormWire, 2.0mm dia.
Total Metal Impurities≤0.005%
Indicator
RUO – Research Use Only