Bromothymol Blue, Pure, Conform to ACS
Bromothymol Blue, Pure, Conform to ACS
Bromothymol Blue, Pure, Conform to ACS
Bromothymol Blue, Pure, Conform to ACS
Thermo Scientific Chemicals
Bromothymol Blue, Pure, Conform to ACS
CAS: 76-59-5 | C27H28Br2O5S | 624.384 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA403251000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBromothymol Blue
Melting Point204.0°C
Name NotePure
CAS76-59-5
ClarityPasses Test
View more
Indicator
RUO – Research Use Only