3-Bromo-1-propanol, 92%, Thermo Scientific Chemicals
3-Bromo-1-propanol, 92%, Thermo Scientific Chemicals
3-Bromo-1-propanol, 92%, Thermo Scientific Chemicals
3-Bromo-1-propanol, 92%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Bromo-1-propanol, 92%, Thermo Scientific Chemicals
CAS: 627-18-9 | C3H7BrO | 138.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA403200250
Price (MYR)
1,292.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,292.00
Add to cart
Chemical Identifiers
CAS7533-40-6
IUPAC Name(2S)-2-amino-4-methylpentan-1-ol
Molecular FormulaC6H15NO
InChI KeyVPSSPAXIFBTOHY-LURJTMIESA-N
SMILESCC(C)C[C@H](N)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Viscous liquid
Infrared spectrumConforms
GC>=97.5 %
Specific optical rotation+3° to +5° (20°C, 589 nm) (c=9, ethanol)