Bromophenol Blue, pure, 0.1% in water, Thermo Scientific Chemicals
Bromophenol Blue, pure, 0.1% in water, Thermo Scientific Chemicals
Bromophenol Blue, pure, 0.1% in water, Thermo Scientific Chemicals
Bromophenol Blue, pure, 0.1% in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bromophenol Blue, pure, 0.1% in water, Thermo Scientific Chemicals
CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Catalog number FSA403151000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS115-39-9, 7732-18-5
Specifications
Chemical Name or MaterialBromophenol Blue
CAS115-39-9
CAS7732-18-5
Concentrationabout 0.1% (by UV)
Molecular FormulaC19H10Br4O5S
View more
Indicator
RUO – Research Use Only