2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromo-2-methylpropionyl Bromide, 98%, Thermo Scientific Chemicals
CAS: 20769-85-1 | C4H6Br2O | 229.9 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA403091000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7681-11-0
IUPAC Namepotassium iodide
Molecular FormulaIK
InChI KeyNLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES[K+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Thiosulphate (S2O3)=<100 ppm
Heavy metals (as Pb)=<10 ppm
Iodate (IO3)=<4 ppm
Appearance (Color)White
Titration Argentometric98.5 to 101.5 %