Bromine, 99+%, extra pure, Thermo Scientific Chemicals
Bromine, 99+%, extra pure, Thermo Scientific Chemicals
Bromine, 99+%, extra pure, Thermo Scientific Chemicals
Bromine, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bromine, 99+%, extra pure, Thermo Scientific Chemicals
CAS: 7726-95-6 | Br | 79.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
1 L
Packaging:
Plastic Coated Glass Bottle
Catalog number FSA402845000
Price (MYR)
1,843.00
Add to cart
Quantity:
500 mL
Packaging:
Plastic Coated Glass Bottle
Request bulk or custom format
Price (MYR)
1,843.00
Add to cart
Chemical Identifiers
CAS24677-78-9
IUPAC Name2,3-dihydroxybenzaldehyde
Molecular FormulaC7H6O3
InChI KeyIXWOUPGDGMCKGT-UHFFFAOYSA-N
SMILESOC1=CC=CC(C=O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to green-beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point104°C to 109°C
Titration with NaOH>=96.0 %