Biebrich Scarlet, dye content approx. 70%
Biebrich Scarlet, dye content approx. 70%
Biebrich Scarlet, dye content approx. 70%
Biebrich Scarlet, dye content approx. 70%
Thermo Scientific Chemicals
Biebrich Scarlet, dye content approx. 70%
CAS: 4196-99-0 | C22H14N4Na2O7S2 | 556.47 g/mol
Have Questions?
Catalog NumberQuantity
FSA402221000100 g
Catalog number FSA402221000
Price (MYR)
519.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
519.00
Add to cart
Chemical Identifiers
CAS302-72-7
IUPAC Name2-aminopropanoic acid
Molecular FormulaC3H7NO2
InChI KeyQNAYBMKLOCPYGJ-UHFFFAOYNA-N
SMILESCC(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Loss on drying=<0.2 % (105°C, 3 hrs)
Chloride (Cl)=<200 ppm
Iron (Fe)=<20 ppm
Titration with HClO498.5 to 101.5 %