Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals
Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals
Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals
Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzamidine hydrochloride, 99%, Thermo Scientific Chemicals
CAS: 1670-14- | C7H9ClN2 | 156.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA401790250
Price (MYR)
441.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
441.00
Add to cart
Chemical Identifiers
CAS17341-93-4
IUPAC Name2,2,2-trichloroethyl carbonochloridate
Molecular FormulaC3H2Cl4O2
InChI KeyLJCZNYWLQZZIOS-UHFFFAOYSA-N
SMILESClC(=O)OCC(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
Refractive Index1.4680-1.4720 @ 20?C