4,4'-Azodianiline, 95%, Thermo Scientific Chemicals
4,4'-Azodianiline, 95%, Thermo Scientific Chemicals
4,4'-Azodianiline, 95%, Thermo Scientific Chemicals
4,4'-Azodianiline, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4,4'-Azodianiline, 95%, Thermo Scientific Chemicals
CAS: 538-41-0 | C12H12N4 | 212.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA401580050
Price (MYR)
1,613.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,613.00
Add to cart
Chemical Identifiers
CAS13822-56-5
IUPAC Name(3-aminopropyl)trimethoxysilane
Molecular FormulaC6H17NO3Si
InChI KeySJECZPVISLOESU-UHFFFAOYSA-N
SMILESCO[Si](CCCN)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Refractive index1.4220 to 1.4250 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid