Azobenzene, 98%, Thermo Scientific Chemicals
Azobenzene, 98%, Thermo Scientific Chemicals
Azobenzene, 98%, Thermo Scientific Chemicals
Azobenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Azobenzene, 98%, Thermo Scientific Chemicals
CAS: 103-33-3 | C12H10N2 | 182.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA401540500
Price (MYR)
958.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
958.00
Add to cart
Chemical Identifiers
CAS6476-36-4
IUPAC Nametris(propan-2-yl)phosphane
Molecular FormulaC9H21P
InChI KeyIGNTWNVBGLNYDV-UHFFFAOYSA-N
SMILESCC(C)P(C(C)C)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
NMR (31P-NMR)>=97.5 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid