Aniline Blue, sodium salt, pure, water soluble, certified
Aniline Blue, sodium salt, pure, water soluble, certified
Aniline Blue, sodium salt, pure, water soluble, certified
Aniline Blue, sodium salt, pure, water soluble, certified
Thermo Scientific Chemicals
Aniline Blue, sodium salt, pure, water soluble, certified
CAS: 28983-56-4 | C37H26N3Na2O9S3 | 798.79 g/mol
Have Questions?
Catalog NumberQuantity
FSA40118025025 g
Catalog number FSA401180250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAniline Blue, sodium salt
CAS28983-56-4
Health Hazard 1Signal Word: WARNING!
Absorption Ratio0.96 to 1.09
Health Hazard 2GHS H Statement: