Ammonium carbonate, ACS reagent, Thermo Scientific Chemicals
Ammonium carbonate, ACS reagent, Thermo Scientific Chemicals
Ammonium carbonate, ACS reagent, Thermo Scientific Chemicals
Ammonium carbonate, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ammonium carbonate, ACS reagent, Thermo Scientific Chemicals
CAS: 506-87-6 | CH8N2O3 | 96.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
HDPE plastic bottle
Catalog number FSA401130010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
HDPE plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium carbonate
Melting Point58.0°C
Name NoteFor analysis ACS
CAS506-87-6
ColorWhite to Yellow