2-Amino-1,3-propanediol, 98%, Thermo Scientific Chemicals
2-Amino-1,3-propanediol, 98%, Thermo Scientific Chemicals
2-Amino-1,3-propanediol, 98%, Thermo Scientific Chemicals
2-Amino-1,3-propanediol, 98%, Thermo Scientific Chemicals
Alfa Aesar
2-Amino-1,3-propanediol, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
View all
Glass bottle
Catalog number FSA401030010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Amino-1, 3-propanediol
CAS534-03-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0