1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-(2-Aminoethyl)piperazine, 99%, Thermo Scientific Chemicals
CAS: 140-31-8 | C6H15N3 | 129.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA400761000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS687-47-8
IUPAC Nameethyl (2S)-2-hydroxypropanoate
Molecular FormulaC5H10O3
InChI KeyLZCLXQDLBQLTDK-BYPYZUCNSA-N
SMILESCCOC(=O)[C@H](C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<50 APHA
GC>=96.0 %
Refractive index1.4120 to 1.4140 (20°C, 589 nm)
Appearance (Form)Clear liquid
Water=<0.3 % (K.F.)