2'-Aminoacetophenone, 97%, Thermo Scientific Chemicals
2'-Aminoacetophenone, 97%, Thermo Scientific Chemicals
2'-Aminoacetophenone, 97%, Thermo Scientific Chemicals
2'-Aminoacetophenone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2'-Aminoacetophenone, 97%, Thermo Scientific Chemicals
CAS: 551-93-9 | C8H9NO | 135.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
10 g
Packaging:
Glass bottle
Catalog number FSA400620500
Price (MYR)
1,994.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,994.00
Add to cart
Chemical Identifiers
CAS18156-74-6
IUPAC Name1-(trimethylsilyl)-1H-imidazole
Molecular FormulaC6H12N2Si
InChI KeyYKFRUJSEPGHZFJ-UHFFFAOYSA-N
SMILESC[Si](C)(C)N1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Refractive index1.4750 to 1.4780 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Appearance (Form)Liquid