Alizarin Red S, pure, certified
Alizarin Red S, pure, certified
Alizarin Red S, pure, certified
Alizarin Red S, pure, certified
Thermo Scientific Chemicals
Alizarin Red S, pure, certified
CAS: 130-22-3 | C14H7NaO7S | 342.253 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number FSA400481000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS1310-66-3
IUPAC Namelithium(1+) hydrate hydroxide
Molecular FormulaH3LiO2
InChI KeyGLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES[Li+].O.[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HCl>=98.0 %
Heavy metals (ICP-OES)=<0.002 %
Sulfate (SO4)=<0.05 %
Lithium carbonate=<2.0 %
View more
Indicator
RUO – Research Use Only