Alcian Blue 8GX, certified
Alcian Blue 8GX, certified
Alcian Blue 8GX, certified
Alcian Blue 8GX, certified
Thermo Scientific Chemicals
Alcian Blue 8GX, certified
CAS: 33864-99-2 | C56H68Cl4CuN16S4 | 1298.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA400461000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAlcian Blue 8GX
Melting Point148.0°C
CAS33864-99-2
Dye Content50% min.
Molecular FormulaC56H68Cl4CuN16S4