Acetylcholine iodide, 99%, Thermo Scientific Chemicals
Acetylcholine iodide, 99%, Thermo Scientific Chemicals
Acetylcholine iodide, 99%, Thermo Scientific Chemicals
Acetylcholine iodide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetylcholine iodide, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number FSA400180100
Price (MYR)
818.00
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
818.00
Add to cart
Specifications
Chemical Name or MaterialAcetylcholine iodide
Name Note99%
CAS2260-50-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning