3-Nitrosalicylic acid, 98%, Thermo Scientific Chemicals
3-Nitrosalicylic acid, 98%, Thermo Scientific Chemicals
3-Nitrosalicylic acid, 98%, Thermo Scientific Chemicals
3-Nitrosalicylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Nitrosalicylic acid, 98%, Thermo Scientific Chemicals
CAS: 85-38-1 | C7H5NO5 | 183.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA399820010
Price (MYR)
336.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
336.00
Add to cart
Chemical Identifiers
CAS1569-02-4
IUPAC Name1-ethoxypropan-2-ol
Molecular FormulaC5H12O2
InChI KeyJOLQKTGDSGKSKJ-UHFFFAOYNA-N
SMILESCCOCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Water=<0.1 % (K.F.)
Appearance (Form)Liquid
GC>=94 % Remark: balance 2-ethoxy-1-propanol
Refractive index1.4050 to 1.4090 (20°C, 589 nm)