5-Methoxytryptamine, 97%
5-Methoxytryptamine, 97%
5-Methoxytryptamine, 97%
5-Methoxytryptamine, 97%
Thermo Scientific Chemicals
5-Methoxytryptamine, 97%
CAS: 608-07-1 | C11H14N2O | 190.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA3985800101 gGlass bottle
Catalog number FSA398580010
Price (MYR)
753.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
753.00
Add to cart
Chemical Identifiers
CAS64438-53-5
IUPAC Namesamarium(3+) tris(2,4-dioxopentan-3-ide)
Molecular FormulaC15H21O6Sm
InChI KeyJTZJMDRBZZVNSP-UHFFFAOYSA-N
SMILES[Sm+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.