Imidazole-2-carboxaldehyde, 98%, Thermo Scientific Chemicals
Imidazole-2-carboxaldehyde, 98%, Thermo Scientific Chemicals
Imidazole-2-carboxaldehyde, 98%, Thermo Scientific Chemicals
Imidazole-2-carboxaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Imidazole-2-carboxaldehyde, 98%, Thermo Scientific Chemicals
CAS: 10111-08-7 | C4H4N2O | 96.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Catalog number FSA398470010
Price (MYR)
869.00
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (MYR)
869.00
Add to cart
Chemical Identifiers
CAS55-55-0
IUPAC Namebis(4-(methylamino)phenol); sulfuric acid
Molecular FormulaC14H20N2O6S
InChI KeyZVNPWFOVUDMGRP-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.CNC1=CC=C(O)C=C1.CNC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Suitability(for determ. of phosphate) Passes test
Appearance (Form)Crystalline powder
Titration99.0 to 101.5 %
Residue after ignition=<0.1 %
Appearance (Color)White to off-white