Glass beads, 400-800 micron, Thermo Scientific Chemicals
Glass beads, 400-800 micron, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glass beads, 400-800 micron, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number FSA397641000
Price (MYR)
443.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
443.00
Add to cart
Chemical Identifiers
CAS106-42-3
IUPAC Name1,4-xylene
Molecular FormulaC8H10
InChI KeyURLKBWYHVLBVBO-UHFFFAOYSA-N
SMILESCC1=CC=C(C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.0 %
Refractive index1.4940 to 1.4960 (20°C, 589 nm)
Residue after evaporation=<0.005 %
Color scale=<10 APHA