Glass beads, 400-800 micron, Thermo Scientific Chemicals
Glass beads, 400-800 micron, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Glass beads, 400-800 micron, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number FSA397640250
Price (MYR)
290.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
290.00
Add to cart
Chemical Identifiers
CAS13574-13-5
IUPAC Name(2S)-5-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoate
Molecular FormulaC17H22NO6
InChI KeyAJDUMMXHVCMISJ-ZDUSSCGKSA-M
SMILESCC(C)(C)OC(=O)N[C@@H](CCC(=O)OCC1=CC=CC=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%