4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-(Bromomethyl)pyridine hydrobromide, 97%, Thermo Scientific Chemicals
CAS: 73870-24-3 | C6H7Br2N | 252.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number FSA397330050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS1702-17-6
IUPAC Name3,6-dichloropyridine-2-carboxylic acid
Molecular FormulaC6H3Cl2NO2
InChI KeyHUBANNPOLNYSAD-UHFFFAOYSA-N
SMILESOC(=O)C1=NC(Cl)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Aqueous acid-base Titration)≥95.0 to ≤105.0%
Assay (Silylated GC)≥95.0%
Appearance (Color)White