Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Castor oil, ethoxylated, pH-range 6.0-8.1, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
500 g
2.5 kg
Catalog number FSA397280010
Price (MYR)
951.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
951.00
Add to cart
Chemical Identifiers
CAS109-86-4
IUPAC Name2-methoxyethan-1-ol
Molecular FormulaC3H8O2
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.2 %
Refractive index1.4010 to 1.4040 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<15 APHA